Orders LinkContactUsLinkComponent
Electronic crimping tools
Electronic crimping tools
Electronic decapper for 20 mm aluminium crimp caps
Katalog # MANA735620
Leverandør:   MACHEREY-NAGEL
Electronic crimping tools
Katalog # MANA735620
Leverandør:  MACHEREY-NAGEL
Electronic crimping tools
Electronic crimping tools
Electronic decapper for 20 mm aluminium crimp caps
Price
10.190,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These battery-powered electronic crimping tools are suitable for aluminium crimp and Flip Top / Flip Off caps (not suitable for 20 mm magnetic / bi-metal crimp caps).
  • Mobile tools for consistent and reproducible crimping results
  • Crimping pressure adjustable by the control unit on top of the tool
  • Long lasting lithium ion cell batteries (full battery charge for several hundred vials, life time of battery >1500 charges)
Certifications: CE certificate of conformity along with one year warranty.
Specifikationer
  • Beskrivelse:
    Electronic decapper for 20 mm aluminium crimp caps
  • Seq:
    08
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rack for 2 electronic crimping tools
    Rack for 2 electronic crimping tools
    MANA735509
    Log ind for tilgængelighed
    1 * 1 Items - 209,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement battery 6,6 V, 8,6 Wh
    Replacement battery 6,6 V, 8,6 Wh
    MANA735500
    Log ind for tilgængelighed
    1 * 1 Items - 1.500,00 kr