VISOCOLOR® ECO Fish analysis case
VISOCOLOR® FISH reagent case manual, 1 manual, 270×180×3 mm (Including packaging), Hazardous material: No, 1
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
The VISOCOLOR Fish analysis kit for water quality testing.
- Specially developed to meet the requirements for testing fishing waters
It combines reagents and accessories for the 6 most important water parameters: ammonium, nitrate, nitrite, phosphate, pH and total hardness.
Specifikationer
Specifikationer
- Pakn:1
- Beskrivelse:VISOCOLOR® FISH reagent case manual
- Dimensioner:270×180×3 mm (Including packaging)
- Model:Hazardous material: No
- Indhold:1 manual
- Cat. No.:MANA933151
- Platform:Accessories
- Holdbarhed (måneder):—
- Vægt:69,9 g (Including packaging)
- Model:MANA933151
- Opbevaringstemperatur:—
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



