VWR® Basic, Overalls, with Hood
blå, XXL
Katalog # 113-9036
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These disposable overalls, with hood, are made from non woven PE-laminated, PP fabric.
- Hood
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Emballering: 1 piece per bag, 50 bags per case.
Documents
Specifikationer
- Tøj størrelse:XXL
- Farve:blå
- Seq:11
- Cat. No.:113-9036
- Vægt:40 g/m²
- Outfit applikationer:General Purpose|Laboratory