VWR® Basic, Overalls, with Hood
blå, XL
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with hood, are made from non woven PE-laminated, PP fabric.
- Hood
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Specifications
- Tøj størrelse:XL
- Farve:blå
- Seq:10
- Cat. No.:113-9035
- Vægt:40 g/m²
- Outfit applikationer:General Purpose|Laboratory