VWR® Basic, Overalls, with Hood
blå, M
Katalog # 113-9021
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Hood
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Emballering: 1 piece per bag, 50 bags per case.
Documents
Specifikationer
- Tøj størrelse:M
- Farve:blå
- Seq:08
- Cat. No.:113-9021
- Vægt:40 g/m²
- Outfit applikationer:General Purpose|Laboratory