VWR® Basic, Overalls, Dual-Zip Closure
hvid, L
Katalog # 113-9011
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (dual-direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Emballering: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Yderligere produktdetaljer
VWR® Non Regulated Disposables for Production
Produktlitteratur
Specifikationer
- Tøj størrelse:L
- Farve:hvid
- Seq:02
- Cat. No.:113-9011
- Vægt:40 g/m²
- Outfit applikationer:General Purpose|Laboratory|Industrial
- Lukkestørrelse:Med lynlås