VWR® Basic Visitor Coats, Velcro® Closure
blå, XS, 92 cm
Katalog # 113-8963
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These disposable coats, with collar, are made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- VELCRO® closure
- Elastic wrists
- Free from natural rubber latex
- Available in white and blue
Weight: 30 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Specifikationer
- Tøj størrelse:XS
- Farve:blå
- Længde:92 cm
- Seq:08
- Køn:Unisex
- Cat. No.:113-8963
- Vægt:30 g/m²
- Outfit applikationer:Laboratory
- Lukkestørrelse:Med Velcro®
- Lommer:uden lommer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



