Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Threaded fittings

Container connection, PP, 25,4 mm, NW 24 mm, white, PP, 25,4 mm
Catalog # BURK8533-0100
Supplier:   BURKLE
Threaded fittings
Catalog # BURK8533-0100
Supplier:  BURKLE
Threaded fittings
Threaded fittings

Container connection, PP, 25,4 mm, NW 24 mm, white, PP, 25,4 mm

Price
158,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Gevind:
    25,4 mm
  • Beskrivelse:
    Container connection, PP, 25,4 mm, NW 24 mm, white
  • Materiale:
    PP
  • Seq:
    31
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.