VWR®, IOS W-Plan Objectives
IOS W-Plan objective, 20×/0,40
Katalog # 630-2951
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
- Ensure field flatness up to F.N. 22
- Based on infinity-corrected optical system
- 100× is also available with iris for darkfield
Documents
VWR® for Microscopy
Produktlitteratur
Specifikationer
- Forstørrelse:20×/0,40
- Beskrivelse:IOS W-Plan objective
- Seq:153