Orders LinkContactUsLinkComponent
VWR®, IOS W-Plan Objectives
VWR®, IOS W-Plan Objectives
IOS W-Plan objective, 2×/0,05
Katalog # 630-2948
Leverandør:   VWR Collection
VWR®, IOS W-Plan Objectives
Katalog # 630-2948
Leverandør:  VWR Collection
VWR®, IOS W-Plan Objectives
IOS W-Plan objective, 2×/0,05
Price
821,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
  • Ensure field flatness up to F.N. 22
  • Based on infinity-corrected optical system
  • 100× is also available with iris for darkfield
Documents
Specifikationer
  • Forstørrelse:
    2×/0,05
  • Beskrivelse:
    IOS W-Plan objective
  • Seq:
    150