Inspection date labels
Inspection date labels, Vinyl, 2027, gul, 15 mm
Katalog # BRDY834115
Leverandør: Brady
Gå til:
Product Details & Documents
These inspection date labels are made of vinyl cloth. They can be used wherever a frequent and regular inspection of machinery and equipment is required. Simply mark the label with a hole punch and apply wherever you need it.
- Show evidence of unauthorised removal
Use the yearly colour system to easily identify when your equipment was last inspected.
Documents
Specifikationer
- Farve:gul
- Beskrivelse:Inspection date labels
- Dimensioner:15 mm
- Materiale:Vinyl
- Legend:2027