Orders LinkContactUsLinkComponent
LysoView™ Lysosomal Stain
LysoView™ Lysosomal Stain
LysoView™ 405, 1000X in DMSO, 10 µl
Katalog # 70066-T
Leverandør:   Biotium
LysoView™ Lysosomal Stain
Katalog # 70066-T
Leverandør:  Biotium
CAS nummer:  
LysoView™ Lysosomal Stain
LysoView™ Lysosomal Stain
LysoView™ 405, 1000X in DMSO, 10 µl
CAS nummer:  
Price
546,00 kr
Log ind for tilgængelighed
1 * 10 µl
Product Details & Documents
LysoView™ dyes label lysosomes in live cells, and are available with blue, green, visible red, and far-red fluorescence.
  • No-wash, fluorescent staining of lysosomes in live cells
  • Non-toxic for real-time live-cell imaging
  • Lower background than LyosoTracker® dyes
  • Dye options for super-resolution imaging
  • Blue, green, red, or far-red fluorescence
LysoView™ dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step. LysoView™ 540 and LysoView™ 633 exhibit pH-sensitive fluorescence, further enhancing the specificity of staining. LysoView™ 488 has been validated in super-resolution imaging by SIM, while LysoView™ 650 fluorophore is compatible with super-resolution imaging by SIM and STED.
LysoView™ 540 and LysoView™ 633 stain vacuoles in yeast. Staining of bacteria varies between dyes.
Documents
Specifikationer
Specifikationer
  • Pakn:
    10 µl
  • Beskrivelse:
    LysoView™ 405, 1000X in DMSO
Specifikationer