Orders LinkContactUsLinkComponent
High content cell imaging system, ImageXpress Micro 4
Filter set, Cy5
Katalog # MLDV5037556
Leverandør:   Molecular Devices
High content cell imaging system, ImageXpress Micro 4
Katalog # MLDV5037556
Leverandør:  Molecular Devices
High content cell imaging system, ImageXpress Micro 4
High content cell imaging system, ImageXpress Micro 4
Filter set, Cy5
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The ImageXpress Micro 4 high content imaging system represents the culmination of four generations of imaging expertise. The agile design allows you to boost your research with this faster than ever system, while providing the option to upgrade to confocal in the future to align with your research needs.
  • Tackle a wide range of assays with oil and air objectives ranging from 1× to 100×
  • Automated five-position filter cube changer to facilitate multiplex assays with up to five filter sets and four objectives for a single experiment
  • Optically-encoded linear voice coil driven stages with encoders that measure position with better than 25 nm resolution, to achieve high accuracy and precision in X, Y and Z axes
  • Expand your research capabilities with environmental control: Temperature, humidity and CO₂ control for multi-day, live cell experiments), transmitted light true phase contrast, confocal imaging (field upgradable), and fluidics (onboard disposable pipetting)
Capture images of whole organisms and cellular or intracellular events with the ImageXpress Micro 4 system, configured to suit your specific biological needs. We offer unlimited configurability with user exchangeable filter cubes, a wide range of objective lenses and environmental control, transmitted light, confocal imaging and fluidic options. Speed up your research by acquiring more than 245000 wells per day at 4× magnification with the system’s unique multi-well crop feature, and resolve the fine detail you need with increasing magnifications.
Combined with MetaXpress high content image acquisition and analysis software, the ImageXpress Micro 4 system is an end-to-end solution to interpret images, understand data and explore ideas.
Specifikationer
  • Beskrivelse:
    Filter set, Cy5
  • Seq:
    107
Complementary Products
Produktinformation
  • Katalog #
    Pakn
    Beskrivelse
    Sterilitet
    Serum format
    Anbefalet arbejdsvolumen
    Steril
    Dyrkningsområde
    Pakket
    Farve
    Arbejdsvolumen
    Tilgængelighed
    Price
  • 30026.
    1 KIT
    Calcein-AM Cell Viability Assay kit, 1000 assays
    Log ind for tilgængelighed
    1 * 1 Each - 3.130,00 kr
  • 30029.
    1 KIT
    NucView™ 488 caspase-3-assay-sæt til levende celler, 100 assays
    Log ind for tilgængelighed
    1 * 1 Each - 4.730,00 kr
  • S1810-500
    Foetal bovine serum, South American origin
    Sterile filtered
    15
    Log ind for tilgængelighed
    1 * 500 mL - 2.400,00 kr
  • 100.103.010
    1
    GrowDex®, 10 ml syringe
    Log ind for tilgængelighed
    1 * 1 Items - 2.170,00 kr
  • 30002.
    1 KIT
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    Log ind for tilgængelighed
    1 * 1 Each - 3.300,00 kr
  • 30009-2
    1 KIT
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    Log ind for tilgængelighed
    1 * 1 Each - 2.210,00 kr
  • 734-2327
    96-brønds plader, flad bund, TC-behandlede
    0,075 - 0,20 ml
    0,33 cm²
    1/pose
    klar
    17
    Log ind for tilgængelighed
    1 * 100 Items - 1.940,00 kr
  • 735-0523
    Microlite™ 1+ strip, 1×12 pr. strip
    Log ind for tilgængelighed
    1 * 320 Items - 1.710,00 kr
  • CORN3904BC
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    +
    75 - 200 µl
    sort
    21
    360 µl
    Log ind for tilgængelighed
    1 * 100 Items - 9.690,00 kr
  • 734-2097
    96 MicroWell™-plade, fladbundede brønde, med låg
    200 µl
    06
    Log ind for tilgængelighed
    1 * 50 Items - 1.630,00 kr