High content cell imaging system, micro confocal, ImageXpress
Product Details & Documents
The ImageXpress® Micro-confocal high content imaging system helps to expand the boundaries of research with a combination of speed, sensitivity and flexibility, giving you higher quality images, at a faster throughput, with more powerful analysis.
- Quickly and easily switch between widefield and several confocal disc configurations, including an exclusive slit option, to capture images at high throughput speeds
- Image more than 160000 wells per day in confocal mode
- Excellent image quality via the unique optical path technology
- Large field of view enables whole-well imaging
- Tackle a wide range of assays with water, oil and air objectives ranging from 1× to 100×
- Expand your research capabilities with water immersion objectives, transmitted light, phase contrast optics, on-board liquid handling, and environmental control options
The ImageXpress® Micro-confocal system lets you capture research quality images of a vast range of assays, including whole organisms, thick tissues, 3-D spheroid assays, and cellular or intracellular events - at a speed you would only expect from widefield screening. Benefit from a wide range of imaging modes, including phase contrast and brightfield label-free imaging, fluorescence, widefield, colorimetric, and confocal imaging, to generate publication quality images at high throughput. The system is designed to be exceptionally versatile with user exchangeable filter cubes, a selection of confocal geometries to match your exact assay requirements, on board fluidics and environmental control, the option to benefit from laser illumination, and much more.
Combined with MetaXpress® high content image acquisition and analysis software, the ImageXpress® Micro-confocal system provides you with a complete, customisable, high throughput screening solution to discover your landmark scientific breakthrough.
Delivery information: Supplied with a 10× plan-apochromatic 0,45 N.A objective, MetaXpress® imaging and analysis software, Windows® 7 64-bit acquisition computer with license to run MetaXpress® powercore and MDCstore database. Confocal screening option includes eight position filter wheel, five position dichroic wheel and dual disk unit.
Documents
3D Analysis and Morphometric Characterization of Compound Effects on Cancer Spheroid Cultures High-content screening of neuronal toxicity using iPSC-derived human neurons High-throughput imaging assays using zebrafish, a model organism for human disease Intelligent time-lapse imaging to enhance live cell assay development Monitoring BNP Expression and Cell Size to Determine Hypertrophic Response in Human Cardiomyocytes
High Content 3D Toxicity Assay using iPSC Derived Hepatocyte Spheroids High Content Assay for Morphological Characterization of 3D Neuronal Networks-in-a microfluidic Platform High Content Assay for Morphological Characterization of Neuronal Develop
Application Notes
ImageXpress® Imaging Solutions
Additional Product Details
Specifikationer
- Beskrivelse:Micro confocal transmitted light option with phase contrast includes 20× Ph1 S plan fluor ELWD adm 0,45 N.A objective
- Seq:06
Complementary Products
Produktinformation
- Katalog #PaknBeskrivelseSterilitetSerum formatSterilPakketArbejdsvolumenFarveBrøndvolumenAnbefalet arbejdsvolumenTilgængelighedPrice
- 30009-21 KITCaspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 mlLog ind for tilgængelighed1 * 1 Each - 2.210,00 kr
- 30026.1 KITCalcein-AM Cell Viability Assay kit, 1000 assaysLog ind for tilgængelighed1 * 1 Each - 3.130,00 kr
- S1810-500Foetal bovine serum, South American originSterile filtered15Log ind for tilgængelighed1 * 500 mL - 2.400,00 kr
- 30002.1 KITViability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assaysLog ind for tilgængelighed1 * 1 Each - 3.300,00 kr
- 30029.1 KITNucView™ 488 caspase-3-assay-sæt til levende celler, 100 assaysLog ind for tilgængelighed1 * 1 Each - 4.730,00 kr
- CORN3904BC96-well microplate, clear flat bottom, tissue culture treated, with lid+75 - 200 µlsort21360 µlLog ind for tilgængelighed1 * 100 Items - 9.690,00 kr
- 734-232796-brønds plader, flad bund, TC-behandlede0,075 - 0,20 ml0,33 cm²1/poseklar17Log ind for tilgængelighed1 * 100 Items - 1.940,00 kr
- 734-209796 MicroWell™-plade, fladbundede brønde, med låg200 µl06Log ind for tilgængelighed1 * 50 Items - 1.630,00 kr