Orders LinkContactUsLinkComponent
High content cell imaging system, micro confocal, ImageXpress
Confocal screening option, dual disk unit allows selection between 3 modes a) 60 micron pinhole disk for your basic confocal requirements b) 50 micron slit for high throughput confocal requirements and c) wide field (not-confocal) imaging mode
  MLDV5039731
High content cell imaging system, micro confocal, ImageXpress
  MLDV5039731
High content cell imaging system, micro confocal, ImageXpress
High content cell imaging system, micro confocal, ImageXpress
Confocal screening option, dual disk unit allows selection between 3 modes a) 60 micron pinhole disk for your basic confocal requirements b) 50 micron slit for high throughput confocal requirements and c) wide field (not-confocal) imaging mode
1 * 1 Items
The ImageXpress® Micro-confocal high content imaging system helps to expand the boundaries of research with a combination of speed, sensitivity and flexibility, giving you higher quality images, at a faster throughput, with more powerful analysis.
  • Quickly and easily switch between widefield and several confocal disc configurations, including an exclusive slit option, to capture images at high throughput speeds
  • Image more than 160000 wells per day in confocal mode
  • Excellent image quality via the unique optical path technology
  • Large field of view enables whole-well imaging
  • Tackle a wide range of assays with water, oil and air objectives ranging from 1× to 100×
  • Expand your research capabilities with water immersion objectives, transmitted light, phase contrast optics, on-board liquid handling, and environmental control options
The ImageXpress® Micro-confocal system lets you capture research quality images of a vast range of assays, including whole organisms, thick tissues, 3-D spheroid assays, and cellular or intracellular events - at a speed you would only expect from widefield screening. Benefit from a wide range of imaging modes, including phase contrast and brightfield label-free imaging, fluorescence, widefield, colorimetric, and confocal imaging, to generate publication quality images at high throughput. The system is designed to be exceptionally versatile with user exchangeable filter cubes, a selection of confocal geometries to match your exact assay requirements, on board fluidics and environmental control, the option to benefit from laser illumination, and much more.
Combined with MetaXpress® high content image acquisition and analysis software, the ImageXpress® Micro-confocal system provides you with a complete, customisable, high throughput screening solution to discover your landmark scientific breakthrough.
 : Supplied with a 10× plan-apochromatic 0,45 N.A objective, MetaXpress® imaging and analysis software, Windows® 7 64-bit acquisition computer with license to run MetaXpress® powercore and MDCstore database. Confocal screening option includes eight position filter wheel, five position dichroic wheel and dual disk unit.
  • Beskrivelse:
    Confocal screening option, dual disk unit allows selection between 3 modes a) 60 micron pinhole disk for your basic confocal requirements b) 50 micron slit for high throughput confocal requirements and c) wide field (not-confocal) imaging mode
  • Seq:
    04
Complementary Products
  •  
    Pakn
    Beskrivelse
    Sterilitet
    Serum format
    Steril
    Pakket
    Arbejdsvolumen
    Farve
    Brøndvolumen
    Anbefalet arbejdsvolumen
  • 30009-2
    1 KIT
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 * 1 Each - 2.210,00 kr
  • 30026.
    1 KIT
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 * 1 Each - 3.130,00 kr
  • 100.103.010
    1
    GrowDex®, 10 ml syringe
    1 * 1 Items - 2.170,00 kr
  • S1810-500
    Foetal bovine serum, South American origin
    Sterile filtered
    15
    1 * 500 mL - 2.400,00 kr
  • 30002.
    1 KIT
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    1 * 1 Each - 3.300,00 kr
  • 30029.
    1 KIT
    NucView™ 488 caspase-3-assay-sæt til levende celler, 100 assays
    1 * 1 Each - 4.730,00 kr
  • 735-0523
    Microlite™ 1+ strip, 1×12 pr. strip
    1 * 320 Items - 1.710,00 kr
  • CORN3904BC
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    +
    75 - 200 µl
    sort
    21
    360 µl
    1 * 100 Items - 9.690,00 kr
  • 734-2327
    96-brønds plader, flad bund, TC-behandlede
    0,075 - 0,20 ml
    0,33 cm²
    1/pose
    klar
    17
    1 * 100 Items - 1.940,00 kr
  • 734-2097
    96 MicroWell™-plade, fladbundede brønde, med låg
    200 µl
    06
    1 * 50 Items - 1.630,00 kr