ASTM precision thermometers
ASTM precision thermometer, ASTM 97 F, 0...+120 °F, 1,0 °F, 302 mm, Red special liquid, Total, 6,0 - 7,0 mm
Katalog # AMARA301350-FL
Leverandør: Amarell
Gå til:
Product Details & Documents
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
Certifications: Compliant with ASTM.
Documents
AMARA301860-FL_TDS_EN
Datablad
Specifikationer
- Ø:6,0 - 7,0 mm
- Neds.- dybde:Total
- Beskrivelse:ASTM precision thermometer, ASTM 97 F
- Længde:302 mm
- Seq:95
- Inddeling:1,0 °F
- Påfyldning:Red special liquid
- Område:0...+120 °F