ASTM precision thermometers
ASTM precision thermometer, ASTM 88 F, +50...+392 °F, 2,0 °F, 287 mm, Red special liquid, 57 mm, 6,0 - 7,0 mm
AMARA301260-FL
: Amarell
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
: Compliant with ASTM.
- Ø:6,0 - 7,0 mm
- Neds.- dybde:57 mm
- Beskrivelse:ASTM precision thermometer, ASTM 88 F
- Længde:287 mm
- Seq:85
- Inddeling:2,0 °F
- Påfyldning:Red special liquid
- Område:+50...+392 °F