ASTM precision thermometers
ASTM precision thermometer, ASTM 86 C, +95...+175 °C, 1,0 °C, 167 mm, Red special liquid, 35 mm, 6,0 - 7,0 mm
AMARA301210-FL
: Amarell
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
: Compliant with ASTM.
- Ø:6,0 - 7,0 mm
- Neds.- dybde:35 mm
- Beskrivelse:ASTM precision thermometer, ASTM 86 C
- Længde:167 mm
- Seq:80
- Inddeling:1,0 °C
- Påfyldning:Red special liquid
- Område:+95...+175 °C