ASTM precision thermometers
ASTM precision thermometer, ASTM 83 F, +60...+160 °F, 1,0 °F, 171 mm, Red special liquid, 40 mm, 6,0 - 7,0 mm
Katalog # AMARA301160-FL
Leverandør: Amarell
Gå til:
Product Details & Documents
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
Certifications: Compliant with ASTM.
Documents
AMARA301860-FL_TDS_EN
Datasheet
Specifikationer
- Ø:6,0 - 7,0 mm
- Neds.- dybde:40 mm
- Beskrivelse:ASTM precision thermometer, ASTM 83 F
- Længde:171 mm
- Seq:75
- Inddeling:1,0 °F
- Påfyldning:Red special liquid
- Område:+60...+160 °F