ASTM precision thermometers
ASTM precision thermometer, ASTM 83 C, +15...+70 °C, 1,0 °C, 171 mm, Red special liquid, 40 mm, 6,0 - 7,0 mm
Katalog # AMARA301150-FL
Leverandør: Amarell
Gå til:
Product Details & Documents
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
Certifications: Compliant with ASTM.
Documents
AMARA301860-FL_TDS_EN
Datablad
Specifikationer
- Ø:6,0 - 7,0 mm
- Neds.- dybde:40 mm
- Beskrivelse:ASTM precision thermometer, ASTM 83 C
- Længde:171 mm
- Seq:74
- Inddeling:1,0 °C
- Påfyldning:Red special liquid
- Område:+15...+70 °C