ASTM precision thermometers
ASTM precision thermometer, ASTM 82 F, 0...+220 °F, 2,0 °F, 162 mm, Red special liquid, 30 mm, 6,0 - 7,0 mm
Katalog # AMARA301140-FL
Leverandør: Amarell
Gå til:
Product Details & Documents
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
: Compliant with ASTM.
Specifikationer
- Ø:6,0 - 7,0 mm
- Neds.- dybde:30 mm
- Beskrivelse:ASTM precision thermometer, ASTM 82 F
- Længde:162 mm
- Seq:73
- Inddeling:2,0 °F
- Påfyldning:Red special liquid
- Område:0...+220 °F