ASTM precision thermometers
ASTM precision thermometer, ASTM 82 C, −15...+105 °C, 1,0 °C, 162 mm, Red special liquid, 30 mm, 6,0 - 7,0 mm
Katalog # AMARA301130-FL
Leverandør: Amarell
Gå til:
Product Details & Documents
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
Certifications: Compliant with ASTM.
Documents
AMARA301860-FL_TDS_EN
Datablad
Specifikationer
- Ø:6,0 - 7,0 mm
- Neds.- dybde:30 mm
- Beskrivelse:ASTM precision thermometer, ASTM 82 C
- Længde:162 mm
- Seq:72
- Inddeling:1,0 °C
- Påfyldning:Red special liquid
- Område:−15...+105 °C