ASTM precision thermometers
ASTM precision thermometer, ASTM 5 F, −30...+120 °F, 2,0 °C, 230 mm, Red special liquid, 108 mm, 6,0 - 7,0 mm
Katalog # AMARA300080-FL
Leverandør: Amarell
Gå til:
Product Details & Documents
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
Certifications: Compliant with ASTM.
Documents
AMARA301860-FL_TDS_EN
Datasheet
Specifikationer
- Ø:6,0 - 7,0 mm
- Neds.- dybde:108 mm
- Beskrivelse:ASTM precision thermometer, ASTM 5 F
- Længde:230 mm
- Seq:004
- Inddeling:2,0 °C
- Påfyldning:Red special liquid
- Område:−30...+120 °F