Orders LinkContactUsLinkComponent
Probes for testo 440
Probes for testo 440
16 mm vane probe head incl. temperature probe
245×16×16 mm
Diameter probe head: 16 mm
Length probe shaft: 230 mm, 10...70 °C
V: 0,6 - 50 m/s, T: ±1,8 °C
V: ±(0,2 m/s +1% of mv) (0,6 - 40 m/s); ±(0,2 m/s - 2% of mv) (40,1 - 50 m/s), T: 0,1 °C
V: 0,01 m/s
Catalog # TEST0635.9570
Supplier:   Testo
Probes for testo 440
Catalog # TEST0635.9570
Supplier:  Testo
Probes for testo 440
Probes for testo 440
16 mm vane probe head incl. temperature probe
245×16×16 mm
Diameter probe head: 16 mm
Length probe shaft: 230 mm, 10...70 °C
V: 0,6 - 50 m/s, T: ±1,8 °C
V: ±(0,2 m/s +1% of mv) (0,6 - 40 m/s); ±(0,2 m/s - 2% of mv) (40,1 - 50 m/s), T: 0,1 °C
V: 0,01 m/s
Price
4.920,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.
Specifications
  • Område:
    10...70 °C
    V: 0,6 - 50 m/s
  • Opløsning:
    T: 0,1 °C
    V: 0,01 m/s
  • Beskrivelse:
    16 mm vane probe head incl. temperature probe
    245×16×16 mm
    Diameter probe head: 16 mm
    Length probe shaft: 230 mm
  • Seq:
    01
  • Nøjagtighed:
    T: ±1,8 °C
    V: ±(0,2 m/s +1% of mv) (0,6 - 40 m/s); ±(0,2 m/s - 2% of mv) (40,1 - 50 m/s)