Probes for testo 440
CO₂ probe head, 195×50×40 mm, diameter probe head: 30 mm, fixed cable: 1,4 m, 0 - 500 ppm, CO₂: ±3 ppm (0 - 30 ppm); ±5 ppm (30,1 - 100 ppm); ±10% of mv (100,1 - 500 ppm), 0,1 ppm
TEST0632.1272
: Testo
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.
Documents
- Område:0 - 500 ppm
- Opløsning:0,1 ppm
- Beskrivelse:CO₂ probe head, 195×50×40 mm, diameter probe head: 30 mm, fixed cable: 1,4 m
- Seq:06
- Nøjagtighed:CO₂: ±3 ppm (0 - 30 ppm); ±5 ppm (30,1 - 100 ppm); ±10% of mv (100,1 - 500 ppm)