Orders LinkContactUsLinkComponent
Probes for testo 440
Probes for testo 440
Temperature and humidity probe, 295×50×40 mm, diameter probe head: 12 mm, fixed cable: 1,4 m, −20...+70 °C
RH: 0 - 100 %, T: 0,5 °C
RH: ±2% (5 - 90%); ±0,03% RH/K (k=1), T: 0,1 °C
RH: 0,1 %
  TEST0636.9732
 :   Testo
Probes for testo 440
  TEST0636.9732
 :  Testo
Probes for testo 440
Probes for testo 440
Temperature and humidity probe, 295×50×40 mm, diameter probe head: 12 mm, fixed cable: 1,4 m, −20...+70 °C
RH: 0 - 100 %, T: 0,5 °C
RH: ±2% (5 - 90%); ±0,03% RH/K (k=1), T: 0,1 °C
RH: 0,1 %
1.320,00 kr
1 * 1 Items
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.
  • Område:
    −20...+70 °C
    RH: 0 - 100 %
  • Opløsning:
    T: 0,1 °C
    RH: 0,1 %
  • Beskrivelse:
    Temperature and humidity probe, 295×50×40 mm, diameter probe head: 12 mm, fixed cable: 1,4 m
  • Seq:
    25
  • Nøjagtighed:
    T: 0,5 °C
    RH: ±2% (5 - 90%); ±0,03% RH/K (k=1)