Orders LinkContactUsLinkComponent
Probes for testo 440
Probes for testo 440
Vane probe incl. temperature sensor, 375×105×46 mm, diameter probe head: 100 mm, fixed cable: 1,4 m, −20...+70 °C
V: 0,3 - 35 m/s, T: ±0,5 °C
V: ±(0,1 m/s +1,5% of mv) (0,3 - 20 m/s); ±(0,2 m/s - 1,5% of mv) (20,01 - 35 m/s), T: 0,1 °C
V: 0,01 m/s
Katalog # TEST0635.9432
Leverandør:   Testo
Probes for testo 440
Katalog # TEST0635.9432
Leverandør:  Testo
Probes for testo 440
Probes for testo 440
Vane probe incl. temperature sensor, 375×105×46 mm, diameter probe head: 100 mm, fixed cable: 1,4 m, −20...+70 °C
V: 0,3 - 35 m/s, T: ±0,5 °C
V: ±(0,1 m/s +1,5% of mv) (0,3 - 20 m/s); ±(0,2 m/s - 1,5% of mv) (20,01 - 35 m/s), T: 0,1 °C
V: 0,01 m/s
Price
kr3,590.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.
Specifikationer
  • Område:
    −20...+70 °C
    V: 0,3 - 35 m/s
  • Opløsning:
    T: 0,1 °C
    V: 0,01 m/s
  • Beskrivelse:
    Vane probe incl. temperature sensor, 375×105×46 mm, diameter probe head: 100 mm, fixed cable: 1,4 m
  • Seq:
    28
  • Nøjagtighed:
    T: ±0,5 °C
    V: ±(0,1 m/s +1,5% of mv) (0,3 - 20 m/s); ±(0,2 m/s - 1,5% of mv) (20,01 - 35 m/s)