Warning signs
General warning, Polyester, 50×43 mm
Katalog # BRDY826613
Leverandør: Brady
Gå til:
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Specifikationer
- Beskrivelse:General warning
- Dimensioner:50×43 mm
- Materiale:Polyester
- Seq:07