Orders LinkContactUsLinkComponent
High Stackable Ledges, 100 mm
High Stackable Ledges, 100 mm
Shelf ledge, Metroseal green with Microban®, 610 mm width
Katalog # METOL24N-4K3
Leverandør:   Metro International
High Stackable Ledges, 100 mm
Katalog # METOL24N-4K3
Leverandør:  Metro International
High Stackable Ledges, 100 mm
Shelf ledge, Metroseal green with Microban®, 610 mm width
Price
390,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Actual ledge length is approximately 25 mm shorter than nominal shelf length/width.
Specifikationer
  • Beskrivelse:
    Shelf ledge, Metroseal green with Microban®, 610 mm width
  • Seq:
    03