Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Carbon dioxide standard solutions

Carbon dioxide standard solution, 4 mg/l, 100×2 ml ampoules, 100
Katalog # REGESC024C
Leverandør:   REAGECON
Carbon dioxide standard solutions
Katalog # REGESC024C
Leverandør:  REAGECON
CAS nummer:  
Carbon dioxide standard solutions

Carbon dioxide standard solution, 4 mg/l, 100×2 ml ampoules, 100

CAS nummer:  
Price
3.620,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Standards for M965D carbon dioxide analyser, for soft drinks and brewery applications.

Specifikationer

Specifikationer

  • Koncentration:
    4 mg/l
  • Pakn:
    100
  • Beskrivelse:
    Carbon dioxide standard solution
  • Pk Info:
    100×2 ml ampoules

Specifikationer