Ribbons for label printer BBP™11
Thermal transfer printer ribbon, halogen free, 6000 series, width 65 mm, BBP™11, 70 m, sort
Catalog # BRDY196515
Supplier: Brady
Jump to:
Product Details & Documents
Documents
Specifications
- Farve:sort
- Beskrivelse:Thermal transfer printer ribbon, halogen free, 6000 series, width 65 mm
- Længde:70 m
- Seq:04
- Til:BBP™11