Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sisomicin ≥98%

Sisomicin ≥98%

500 mg
Katalog # CAYM21046-500
Irriterende
Sundhed
Leverandør:   Cayman Chemical
Sisomicin ≥98%
Katalog # CAYM21046-500
Leverandør:  Cayman Chemical
CAS nummer:  32385-11-8
Sisomicin ≥98%

500 mg

CAS nummer:  32385-11-8
Price
2.780,00 kr
Log ind for tilgængelighed
1 * 500 mg
Product Details & Documents

Sisomicin is a broad-spectrum aminoglycoside antibiotic originally isolated from M. inyoensis.

It is active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa among others (MICs = 0.01-0.75 µg/ml) but is less active against P. multocida and V. coli (MICs = 7.5 and 2.4 µg/ml, respectively). Sisomicin is protective against Gram-positive and Gram-negative bacterial infections in mice with 50% protective dose (PD50s) values ranging from 0.5 to 6 mg/kg.

Synonymer:  Rickamicin, Salvamina, SCH 13475, Antibiotic 6640
  • CAS nummer:32385-11-8
  • Formel:C₁₉H₃₇N₅O₇
  • MDL nummer:MFCD00865123
  • Molecular Weight:447.50 g/mol
  • Opbevaringstemperatur:Fryser
  • EINECS:251-018-2
Specifikationer

Specifikationer

  • Pakn:
    500 mg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.