Sisomicin ≥98%
50 mg
CAYM21046-50
Sisomicin is a broad-spectrum aminoglycoside antibiotic originally isolated from M. inyoensis.
It is active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa among others (MICs = 0.01-0.75 µg/ml) but is less active against P. multocida and V. coli (MICs = 7.5 and 2.4 µg/ml, respectively). Sisomicin is protective against Gram-positive and Gram-negative bacterial infections in mice with 50% protective dose (PD50s) values ranging from 0.5 to 6 mg/kg.
: Rickamicin, Salvamina, SCH 13475, Antibiotic 6640
- :32385-11-8
- :C₁₉H₃₇N₅O₇
- :MFCD00865123
- :447.50 g/mol
- :Fryser
- :251-018-2
- Pakn:50 mg