Orders LinkContactUsLinkComponent
Sisomicin ≥98%
50 mg
  CAYM21046-50
Sisomicin ≥98%
  CAYM21046-50
 :  32385-11-8
Sisomicin ≥98%
50 mg
 :  32385-11-8
395,00 kr
1 * 50 mg
Sisomicin is a broad-spectrum aminoglycoside antibiotic originally isolated from M. inyoensis.
It is active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa among others (MICs = 0.01-0.75 µg/ml) but is less active against P. multocida and V. coli (MICs = 7.5 and 2.4 µg/ml, respectively). Sisomicin is protective against Gram-positive and Gram-negative bacterial infections in mice with 50% protective dose (PD50s) values ranging from 0.5 to 6 mg/kg.
 :  Rickamicin, Salvamina, SCH 13475, Antibiotic 6640
  •  :32385-11-8
  •  :C₁₉H₃₇N₅O₇
  •  :MFCD00865123
  •  :447.50 g/mol
  •  :Fryser
  •  :251-018-2
  • Pakn:
    50 mg