Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sisomicin ≥98%

Sisomicin ≥98%

100 mg
Katalog # CAYM21046-100
Irriterende
Sundhed
Leverandør:   Cayman Chemical
Sisomicin ≥98%
Katalog # CAYM21046-100
Leverandør:  Cayman Chemical
CAS nummer:  32385-11-8
Sisomicin ≥98%

100 mg

CAS nummer:  32385-11-8
Price
695,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Sisomicin is a broad-spectrum aminoglycoside antibiotic originally isolated from M. inyoensis.

It is active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa among others (MICs = 0.01-0.75 µg/ml) but is less active against P. multocida and V. coli (MICs = 7.5 and 2.4 µg/ml, respectively). Sisomicin is protective against Gram-positive and Gram-negative bacterial infections in mice with 50% protective dose (PD50s) values ranging from 0.5 to 6 mg/kg.

Synonymer:  Rickamicin, Salvamina, SCH 13475, Antibiotic 6640
  • CAS nummer:32385-11-8
  • Formel:C₁₉H₃₇N₅O₇
  • MDL nummer:MFCD00865123
  • Molecular Weight:447.50 g/mol
  • Opbevaringstemperatur:Fryser
  • EINECS:251-018-2
Specifikationer

Specifikationer

  • Pakn:
    100 mg

Specifikationer