Orders LinkContactUsLinkComponent
Inspection date labels
Inspection date labels
Inspection date labels, Vinyl, 2025, orange, 15 mm
Katalog # BRDY834109
Leverandør:   Brady
Inspection date labels
Katalog # BRDY834109
Leverandør:  Brady
Inspection date labels
Inspection date labels
Inspection date labels, Vinyl, 2025, orange, 15 mm
Price
1.120,00 kr
Log ind for tilgængelighed
1 * 600 Items
Product Details & Documents
These inspection date labels are made of vinyl cloth. They can be used wherever a frequent and regular inspection of machinery and equipment is required. Simply mark the label with a hole punch and apply wherever you need it.
  • Show evidence of unauthorised removal
Use the yearly colour system to easily identify when your equipment was last inspected.
Specifikationer
  • Farve:
    orange
  • Beskrivelse:
    Inspection date labels
  • Dimensioner:
    15 mm
  • Materiale:
    Vinyl
  • Legend:
    2025