Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sheep Recombinant Prl-A

Sheep Recombinant Prl-A

50 µG
Katalog # APOSBITP1255-50UG
Leverandør:   Apollo Scientific
Sheep Recombinant Prl-A
Katalog # APOSBITP1255-50UG
Leverandør:  Apollo Scientific
CAS nummer:  
Sheep Recombinant Prl-A

50 µG

CAS nummer:  
Price
1.530,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Specifikationer

Specifikationer

  • Pakn:
    50 µG
  • Cat. No.:
    APOSBITP1255-50UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Prl-A
  • Protein synonyms:
    Prolactin Antagonist
  • Species:
    Sheep

Specifikationer