Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human recombinant CD72

Human recombinant CD72

10 µG
Katalog # APOSBITP1508-10UG
Leverandør:   Apollo Scientific
Human recombinant CD72
Katalog # APOSBITP1508-10UG
Leverandør:  Apollo Scientific
CAS nummer:  
Human recombinant CD72

10 µG

CAS nummer:  
Price
5.450,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Specifikationer

Specifikationer

  • Pakn:
    10 µG
  • Cat. No.:
    APOSBITP1508-10UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    CD72
  • Species:
    Human

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.