Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
6-Bromisoquinolin

6-Bromisoquinolin

Bottle, 25 g
Katalog # APOSOR14998-25G
Irriterende
Leverandør:   Apollo Scientific
6-Bromisoquinolin
Katalog # APOSOR14998-25G
Leverandør:  Apollo Scientific
CAS nummer:  34784-05-9
6-Bromisoquinolin

Bottle, 25 g

CAS nummer:  34784-05-9
Price
648,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
  • CAS nummer:34784-05-9
  • Formel:C₉H₆BrN
  • MDL nummer:MFCD04973299
  • Molecular Weight:208.06 g/mol
  • Smeltende Pt:42…44 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.