Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
7-Nitroindazol 97%

7-Nitroindazol 97%

Bottle, 5 g
Katalog # APOSOR13550-5G
Giftig
Sundhed
Leverandør:   Apollo Scientific
7-Nitroindazol 97%
Katalog # APOSOR13550-5G
Leverandør:  Apollo Scientific
CAS nummer:  2942-42-9
7-Nitroindazol 97%

Bottle, 5 g

CAS nummer:  2942-42-9
Price
242,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole
  • CAS nummer:2942-42-9
  • Formel:C₇H₅N₃O₂
  • MDL nummer:MFCD00022789
  • Molecular Weight:163.14 g/mol
  • Opbevaringstemperatur:Fryser
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.