Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
ω-Brom-4-hydroxyacetophenon 97%

ω-Brom-4-hydroxyacetophenon 97%

Bottle, 25 g
Katalog # APOSOR11266-25G
Ætsende
Irriterende
Leverandør:   Apollo Scientific
ω-Brom-4-hydroxyacetophenon 97%
Katalog # APOSOR11266-25G
Leverandør:  Apollo Scientific
CAS nummer:  2491-38-5
ω-Brom-4-hydroxyacetophenon 97%

Bottle, 25 g

CAS nummer:  2491-38-5
Price
187,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  2-Bromo-1-(4-hydroxyphenyl)ethanone, 2-Bromo-4'-hydroxyacetophenone, 2-Bromo-1-(4-hydroxyphenyl)ethan-1-one, α-Bromo-4-hydroxyacetophenone, 4-Hydroxyphenacyl bromide
  • CAS nummer:2491-38-5
  • Formel:C₈H₇BrO₂
  • MDL nummer:MFCD00072424
  • Molecular Weight:215.05
  • Opbevaringstemperatur:Køleskab
  • FN:1759
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.