Phenylacetamid 98%
Bottle, 100 g
Katalog # APOSOR0700-100G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Synonymer: 2-Phenylacetamide
- CAS nummer:103-81-1
- Formel:C₈H₉NO
- MDL nummer:MFCD00059193
- Molecular Weight:135.17 g/mol
- Smeltende Pt:156 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-147-0
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:100 g



