Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phenylacetamid 98%

Phenylacetamid 98%

Bottle, 25 g
Catalog # APOSOR0700-25G
Irritant
Supplier:   Apollo Scientific
Phenylacetamid 98%
Catalog # APOSOR0700-25G
Supplier:  Apollo Scientific
CAS Number:  103-81-1
Phenylacetamid 98%

Bottle, 25 g

CAS Number:  103-81-1
Price
165,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Phenylacetamide
  • CAS Number:103-81-1
  • Formula:C₈H₉NO
  • MDL Number:MFCD00059193
  • Molecular Weight:135.17 g/mol
  • Melting Pt:156 °C
  • Storage Temperature:Stue temperatur
  • EINECS:203-147-0
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifications