Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(S)-(-)-2-Acetoxypropionic acid

Bottle, 5 g
Catalog # APOSOR322114-5G
Irritant
Supplier:   Apollo Scientific
(S)-(-)-2-Acetoxypropionic acid
Catalog # APOSOR322114-5G
Supplier:  Apollo Scientific
CAS Number:  6034-46-4
(S)-(-)-2-Acetoxypropionic acid

Bottle, 5 g

CAS Number:  6034-46-4
Price
385,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (-)-O-Acetyl-L-lactic Acid, (S)-2-Acetoxypropanoic acid
  • CAS Number:6034-46-4
  • Formula:C₅H₈O₄
  • MDL Number:MFCD00191370
  • Molecular Weight:132.12 g/mol
  • Boiling Pt:142 °C/20mmHg
  • Density:1,18 g/cm³ (25 °C)
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.