(S)-(-)-2-Acetoxypropionic acid
Bottle, 1 g
Katalog # APOSOR322114-1G
Leverandør: Apollo Scientific
(S)-(-)-2-Acetoxypropionic acid
Katalog # APOSOR322114-1G
Leverandør: Apollo Scientific
CAS nummer: 6034-46-4
Gå til:
Product Details & Documents
Synonymer: (-)-O-Acetyl-L-lactic Acid, (S)-2-Acetoxypropanoic acid
- CAS nummer:6034-46-4
- Formel:C₅H₈O₄
- MDL nummer:MFCD00191370
- Molecular Weight:132.12 g/mol
- Kogende Pt:142 °C/20mmHg
- Tæthed:1,18 g/cm³ (25 °C)
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:1 g



