4'-Demethylepipodophyllotoxin
Bottle, 25 g
Katalog # APOSOR3101T-25G
Leverandør: Apollo Scientific
4'-Demethylepipodophyllotoxin
Katalog # APOSOR3101T-25G
Leverandør: Apollo Scientific
CAS nummer: 6559-91-7
Gå til:
Product Details & Documents
A potent inhibitor of microtubule assembly.
Synonymer: (5R,5aR,8aR,9S)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
- CAS nummer:6559-91-7
- Formel:C₂₁H₂₀O₈
- MDL nummer:MFCD00189421
- Molecular Weight:400.38 g/mol
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:25 g



