Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-Bromethanol 97%

Bottle, 100 g
Katalog # APOSOR30640-100G
Ætsende
Giftig
Leverandør:   Apollo Scientific
2-Bromethanol 97%
Katalog # APOSOR30640-100G
Leverandør:  Apollo Scientific
CAS nummer:  540-51-2
2-Bromethanol 97%

Bottle, 100 g

CAS nummer:  540-51-2
Price
714,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
  • CAS nummer:540-51-2
  • Formel:BrH₂CCH₂OH
  • MDL nummer:MFCD00002827
  • Molecular Weight:124.97 g/mol
  • Kogende Pt:149…150 °C
  • Smeltende Pt:–80 °C
  • Tæthed:1,762 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:208-748-1
  • FN:2922
  • ADR:8+6.1,II
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.