8-Quinolinol
Bottle, 100 g
Katalog # APOSOR2837-100G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Synonymer: 8-Hydroxyquinoline
- CAS nummer:148-24-3
- Formel:C₉H₇NO
- MDL nummer:MFCD00006807
- Molecular Weight:145.16 g/mol
- Kogende Pt:267 °C/252mmHg
- Smeltende Pt:70…76 °C
- Tæthed:1,03 g/cm³ (25 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-711-1
- FN:2811
- ADR:6.1,III
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Bottle
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



