Orders LinkContactUsLinkComponent
BM-2 orchid medium
BM-2 orchid medium 10 L, 10 L
  APOSPMB142N-10L
BM-2 orchid medium
  APOSPMB142N-10L
 :  
BM-2 orchid medium
BM-2 orchid medium 10 L, 10 L
 :  
165,00 kr
1 * 10 L
Contains macro/micronutrients, vitamins and plantgrowth regulators required for orchid culturePlant tissue culture testedFull specification given in appendix.
  • MFCD07370631
  • Pakn:
    10 L
  • Beskrivelse:
    BM-2 orchid medium 10 L