Phenylboronsyre 99%
Bottle, 25 g
Catalog # APOSOR2447-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid
- CAS Number:98-80-6
- Formula:C₆H₇BO₂
- MDL Number:MFCD00002103
- Molecular Weight:121.93 g/mol
- Melting Pt:216…219 °C
- Storage Temperature:Køleskab
- EINECS:202-701-9
Specifications
Specifications
- Pakning:Bottle
- Pakn:25 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



