Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phenylboronsyre 99%

Phenylboronsyre 99%

Bottle, 100 g
Katalog # APOSOR2447-100G
Irriterende
Leverandør:   Apollo Scientific
Phenylboronsyre 99%
Katalog # APOSOR2447-100G
Leverandør:  Apollo Scientific
CAS nummer:  98-80-6
Phenylboronsyre 99%

Bottle, 100 g

CAS nummer:  98-80-6
Price
659,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid
  • CAS nummer:98-80-6
  • Formel:C₆H₇BO₂
  • MDL nummer:MFCD00002103
  • Molecular Weight:121.93 g/mol
  • Smeltende Pt:216…219 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:202-701-9
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.